missing translation for 'onlineSavingsMsg'
Learn More

Tauroursodeoxycholic Acid, TRC

Product Code. 30137970
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137970 5 g 5g
17953475 500 mg 500mg
17954515 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137970 Supplier Toronto Research Chemicals Supplier No. TRCT7793105G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 14605-22-2
Molecular Formula C26 H45 N O6 S
Molecular Weight (g/mol) 499.7
Synonym Ethanesulfonic acid, 2-[[(3α,5β,7β)-3,7-dihydroxy-24-oxocholan-24-yl]amino]-, Taurine, N-(3α,7β-dihydroxy-5β-cholan-24-oyl)- (8CI), Tauroursodeoxycholic acid (6CI,7CI), 2-[[(3α,5β,7β)-3,7-Dihydroxy-24-oxocholan-24-yl]amino]ethanesulfonic acid, Cholane, ethanesulfonic acid deriv., 3α,7β-Dihydroxy-5β-cholanoyltaurine, Taurolite, UR 906, Ursodeoxycholyltaurine
IUPAC Name 2-[[(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonic acid
SMILES C[C@H](CCC(=O)NCCS(=O)(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Tekniske data

Quantity 5 g
InChI Formula InChI=1S/C26H45NO6S/c1-16(4-7-23(30)27-12-13-34(31,32)33)19-5-6-20-24-21(9-11-26(19,20)3)25(2)10-8-18(28)14-17(25)15-22(24)29/h16-22,24,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33)/t16-,17+,18-,19-,20+,21+,22+,24+,25+,26-/m1/s1
Formula Weight 499.2968
Percent Purity >95%
Chemical Name or Material Tauroursodeoxycholic Acid
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.