missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taurodeoxycholic acid sodium hydrate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Taurodeoxycholic acid sodium hydrate (Sodium taurodeoxycholate monohydrate) prevents apoptosis by blocking a calcium-mediated apoptotic pathway as well as caspase-12 activation. Taurodeoxycholic acid sodium hydrate is investigated for use in several conditions such as Primary Biliary Cirrhosis (PBC), insulin resistance, amyloidosis, Cystic Fibrosis, Cholestasis, and Amyotrophic Lateral Sclerosis.
Specifications
Specifications
| Chemical Name or Material | Taurodeoxycholic acid sodium hydrate |
| CAS | 110026-03-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H46NNaO7S |
| Quantity | 10 mg |
| Synonym | Sodium taurodeoxycholate monohydrate |
| Solubility Information | DMSO : 125 mg/mL (231.61 mM; Need ultrasonic) |
| SMILES | C[C@H](CCC(NCCS(=O)(O[Na])=O)=O)[C@H]1CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C.O |
| Molecular Weight (g/mol) | 539.7 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction