missing translation for 'onlineSavingsMsg'
Learn More

Taurine, Tocris Bioscience™

Product Code. 18025232
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18025232 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18025232 Supplier Tocris Bioscience Supplier No. 0209/1G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-selective, endogenous glycine receptor partial agonist

One of the most abundant free amino acids in the brain. A partial agonist at the inhibitory glycine receptor. May be used in protocols for the generation of retinal pigment epithelial cells from hPSCs
TRUSTED_SUSTAINABILITY

Specifications

CAS 107-35-7
Molecular Formula C2H7NO3S
MDL Number MFCD00008197
Quantity 1 g
Synonym taurine, tauphon, ethanesulfonic acid, 2-amino, l-taurine, 2-aminoethylsulfonic acid, o-due, 2-sulfoethylamine, taufon, aminoethanesulfonic acid, aminoethylsulfonic acid
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
SMILES NCCS(O)(=O)=O
IUPAC Name 2-aminoethane-1-sulfonic acid
Molecular Weight (g/mol) 125.14
PubChem CID 1123
ChEBI CHEBI:15891
Formula Weight 125.14
Percent Purity >98%
Chemical Name or Material Taurine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.