missing translation for 'onlineSavingsMsg'
Learn More

Taurine, MedChemExpress

Product Code. p-200097738
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16480888

Brand: Medchem Express HYB0351

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Taurine, a sulphur-containing amino acid and an organic osmolyte involved in cell volume regulation, provides a substrate for the formation of bile salts, and plays a role in the modulation of intracellular free calcium concentration. Taurine has the ability to activate autophagy in adipocytes.

Taurine, a sulphur-containing amino acid and an organic osmolyte involved in cell volume regulation, provides a substrate for the formation of bile salts, and plays a role in the modulation of intracellular free calcium concentration. Taurine has the ability to activate autophagy in adipocytes.

Specifications

Chemical Name or Material Taurine
CAS 107-35-7
Color White
Purity Grade Notes Research
Molecular Formula C2H7NO3S
Quantity 5 g
Synonym 2-Aminoethanesulfonic acid
Solubility Information H2O : 25 mg/mL (199.76 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (7.99 mM; Need ultrasonic)
SMILES NCCS(=O)(O)=O
Molecular Weight (g/mol) 125.15
Formula Weight 125.15
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.