missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau tubulin kinase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-92476
This item is not returnable.
View return policy
Description
Tau tubulin kinase 2 Polyclonal specifically detects Tau tubulin kinase 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Tau tubulin kinase 2 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| EC 2.7.11, EC 2.7.11.1, KIAA0847tau-tubulin kinase 2, SCA11, spinocerebellar ataxia 11, tau tubulin kinase 2, TTBK | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TTBK2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TTEPLDVTKTQTFSVVPNQDKNNEIMKLLTVGTSEISSRDIDPHVEGQIGQVAEMQKNKISKDDDIMSEDLPGHQGDLSTFLHQEGKREKITPRNGELFHCVS | |
| 0.1 mL | |
| Protein Kinase | |
| 146057 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction