missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau tubulin kinase 2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41097DL594
This item is not returnable.
View return policy
Description
Tau tubulin kinase 2 Polyclonal antibody specifically detects Tau tubulin kinase 2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Tau tubulin kinase 2 | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.7.11, EC 2.7.11.1, KIAA0847tau-tubulin kinase 2, SCA11, spinocerebellar ataxia 11, tau tubulin kinase 2, TTBK | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human TTBK2. The immunogen is located within the last 50 amino acids of TTBK2. Amino Acid Squence: ASTKTPQGKSKPASKLSR | |
| 0.1 mL | |
| Protein Kinase | |
| 146057 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction