missing translation for 'onlineSavingsMsg'
Learn More

Tau tubulin kinase 2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93385-0.1ml

 View more versions of this product

Product Code. 18623171

  • £366.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Tau tubulin kinase 2 Polyclonal antibody specifically detects Tau tubulin kinase 2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tau tubulin kinase 2
Polyclonal
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
EC 2.7.11, EC 2.7.11.1, KIAA0847tau-tubulin kinase 2, SCA11, spinocerebellar ataxia 11, tau tubulin kinase 2, TTBK
Recombinant fusion protein containing a sequence corresponding to amino acids 1185-1244 of human TTBK2 (NP_775771.3). LGRSKSPPSHSGSSSSRRSCQQEHCKPSKNGLKGSGSLHHHSASTKTPQGKSKPASKLSR
0.1 mL
Protein Kinase
146057
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt