missing translation for 'onlineSavingsMsg'
Learn More

Tau, Rabbit anti-Human,Mouse,Rat,Sheep, Clone: Tau-5, Novus Biologicals™

Product Code. 18062626 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.20mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18062626 0.025 mg 25µg
18374090 0.2 mg 0.20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18062626 Supplier Novus Biologicals Supplier No. NBP2810910.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Tau Monoclonal specifically detects Tau in Human, Mouse, Rat, Sheep samples. It is validated for Immunocytochemistry/Immunofluorescence, Electron Microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications Immunofluorescence, Immunomicroscopy
Classification Monoclonal
Clone Tau-5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence, Electron Microscopy
Formulation PBS
Gene Alias DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau
Gene Symbols MAPT
Host Species Rabbit
Immunogen Tau protein.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4137
Test Specificity This antibody binds to sequence 210-241 of tau protein.
Target Species Human, Mouse, Rat, Sheep
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.