missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA583934
This item is not returnable.
View return policy
Description
Immunogen sequence: PLEFTFHVEI TPNVQKEQAH SEEHLGRAAF PGAPGEGPEA RGPSLGEDTK EADLPEPSEK QPAAAPRGKP VSRVPQLKAR MVS
Brain tissues from patients with Alzheimer′s are characterized by several histopathological lesions such as neurofibrillary tangles and senile plaques. Neurofibrillary tangles are recognized by anti-Tau while senile plaques are recognized by b-amyloid and ubiquitin antibodies. This antibody localizes Tau proteins along microtubules in axons, somata, dendrites, astrocytes, and on ribosomes (polysomes.).
Specifications
| Tau | |
| Polyclonal | |
| Unconjugated | |
| MAPT | |
| DDPAC, FLJ31424, FTDP-17, MAPTL, MGC138549, MSTD, MTBT1, MTBT2, PPND, PPP1R103, tau | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4137 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P10636 | |
| MAPT | |
| Recombit protein corresponding to Human MAPT | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction