missing translation for 'onlineSavingsMsg'
Learn More

Tau, p Ser324 Antibody (4E8), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen Tau
Clone 4E8
Dilution ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200
Applications ELISA, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30026732
View Documents
Novus Biologicals
NBP3-26409-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30026820
View Documents
Novus Biologicals
NBP3-26409-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tau Monoclonal antibody specifically detects Tau in Human samples. It is validated for ELISA, Immunocytochemistry
Specifications

Specifications

Tau
ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200
Monoclonal
Purified
RUO
Human
DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau
A synthesized peptide derived from Human Tau [p Ser324] [UniProt P10636]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
4E8
ELISA, Immunocytochemistry
Unconjugated
Rabbit
Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
PBS, pH 7.4, 150mM NaCl and 50% glycerol
4137
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.