missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tau Oligomeric Complex Chimeric Recombinant Rabbit Monoclonal Antibody (TOC1)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA551567

Product Code. 30088511

  • £477.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific for amino acids 209-224 of the Tau Oligomeric complex.

The Tau Oligomeric Complex is a protein complex found primarily in the brain. It plays a crucial role in maintaining the structure and stability of microtubules, which are important for cellular transport and neuronal function.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tau Oligomeric Complex Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
Rabbit
Protein A
RUO
Human
Antibody
IgG κ
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
TOC1
Unconjugated
Tau Oligomer; TOC1
The original antibody was generated by immunizing a mouse with electro eluted cross-linked Tau dimers.
200 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.