missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Antibody (Tau-5), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81091F
This item is not returnable.
View return policy
Description
Tau Monoclonal specifically detects Tau in Human, Mouse, Rat, Sheep samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Tau | |
| Monoclonal | |
| FITC | |
| PBS | |
| MAPT | |
| Tau protein. | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers | |
| 4137 | |
| Human, Mouse, Rat, Sheep | |
| Purified |
| Immunofluorescence | |
| Tau-5 | |
| Immunocytochemistry/Immunofluorescence | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody binds to sequence 210-241 of tau protein. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction