missing translation for 'onlineSavingsMsg'
Learn More

Tau Antibody (2E9), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18411136 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411136 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411136 Supplier Novus Biologicals Supplier No. NBP225162JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tau Monoclonal antibody specifically detects Tau in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Proximity Ligation Assay
Classification Monoclonal
Clone 2E9
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau
Host Species Mouse
Immunogen Recombinant full length 441 amino acid human tau. Epitope located in the peptide KDRVQSKIGSLDNITHVPGG, amino acids 347-366.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4137
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.