Learn More
Invitrogen™ TATDN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588351
Description
Immunogen sequence: MSRFKFIDIG INLTDPMFRG IYRGVQKHQD DLQDVIGRAV EIGVKKFMIT GGNLQDSKDA LHLAQTNGMF FSTVGCHPTR CGEFEKNNPD LYLKELLNLA ENNKGKVVAI GECGLDFDRL QFCPKDTQLK YFEKQFELSE QTKLPMFLHC RNSHAEFLDI MKRNRDRCVG GVVHSFDGTK EAAAALIDLD LYIGFNGCSL KTEANLEVLK SIPSEKLMIE TDAPWCGVKS THAGSKYIRT AFPTKKKWES GHCLKDRNEP CHIIQILEIM SAVRDEDPLE LANTLYNNTI KVFFPGI; Positive Samples: LO2, A-549, A-431; Cellular Location: Nucleus.
TATDN1 is a protein coding gene. Putative deoxyribonuclease.
Specifications
| TATDN1 | |
| Polyclonal | |
| Unconjugated | |
| TATDN1 | |
| 2310079P03Rik; CDA11; hepatocarcinoma high expression protein; LOC690068; Putative deoxyribonuclease TATDN1; similar to TatD DNase domain containing 1; TatD DNase domain containing 1; Tatdn1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83940 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.11 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6P1N9 | |
| TATDN1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-297 of human TATDN1 (NP_1144151). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.