missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TATA Element Modulatory Factor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30235
This item is not returnable.
View return policy
Description
TATA Element Modulatory Factor 1 Polyclonal antibody specifically detects TATA Element Modulatory Factor 1 in Human samples. It is validated for Western Blot
Specifications
| TATA Element Modulatory Factor 1 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| TATA element modulatory factor 1 | |
| between 600 and 650 | |
| 20 μg | |
| Signal Transduction | |
| 7110 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction