missing translation for 'onlineSavingsMsg'
Learn More

TATA binding protein TBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£178.00 - £366.00

Specifications

Antigen TATA binding protein TBP
Applications Western Blot, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18333556
View Documents
Novus Biologicals
NBP3-02958-20UL
20 μL
£178.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18373897
View Documents
Novus Biologicals
NBP3-02958-100UL
100 μL
£366.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TATA binding protein TBP Polyclonal specifically detects TATA binding protein TBP in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.
Specifications

Specifications

TATA binding protein TBP
Polyclonal
Purified
RUO
Human, Mouse, Rat
GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TATA binding protein TBP (NP_001165556.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, KnockDown
Unconjugated
Rabbit
Transcription Factors and Regulators
PBS with 50% glycerol, pH7.3.
6908
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.