missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TATA binding protein TBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
TATA binding protein TBP Polyclonal specifically detects TATA binding protein TBP in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.
Specifications
Specifications
| Antigen | TATA binding protein TBP |
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TATA binding protein TBP (NP_001165556.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?