missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TATA binding protein TBP Antibody (C.15200002), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59208
This item is not returnable.
View return policy
Description
TATA binding protein TBP Monoclonal specifically detects TATA binding protein TBP in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Chromatin Immunoprecipitation Sequencing, Knockdown Validated.
Specifications
| TATA binding protein TBP | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| TBP | |
| The exact immunogen is propietary information. | |
| 100 μg | |
| Transcription Factors and Regulators | |
| 6908 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ChIP Assay, KnockDown | |
| C.15200002 | |
| Western Blot 1:500, Chromatin Immunoprecipitation (ChIP) 4-5 ug/IP, Chromatin Immunoprecipitation Sequencing 4 - 5 ug/IP, Knockdown Validated | |
| GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit | |
| Mouse | |
| ammonium sulphate purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction