missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAT complex Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £626.00
Specifications
| Antigen | TAT complex |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17979312
|
Invitrogen™
MA551122 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17958622
![]() |
Invitrogen™
MA551121 |
200 μL |
£626.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Thrombin-antithrombin complex (TAT complex) is a protein complex formed by the binding of thrombin, a serine protease involved in blood clotting, and antithrombin, a serine protease inhibitor that regulates thrombin activity. This complex plays a crucial role in maintaining the balance between blood clotting and preventing excessive clotting, which can lead to thrombosis. The complex is formed when antithrombin binds to thrombin, inhibiting its activity and preventing the formation of blood clots.Specifications
| TAT complex | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| TAT | |
| F2, SERPINC1 | |
| IgG1 κ | |
| Protein A/G | |
| SERPINC1 |
| C1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P00734, P01008 | |
| 2147, 462 | |
| Recombinant Human TAT protein, Ile364-Glu622 + GGGGS*3 + His33-Lys464 (Accession #P00734 & P01008). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title