missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAT Antibody (OTI3G6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | TAT |
|---|---|
| Clone | OTI3G6 |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306293
|
Novus Biologicals
NBP3-45037-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306503
|
Novus Biologicals
NBP3-45037-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAT Monoclonal antibody specifically detects TAT in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| TAT | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.6.1.5, L-tyrosine:2-oxoglutarate aminotransferase, tyrosine aminotransferase, tyrosine aminotransferase, cytosolic | |
| Full length human recombiant protein of human TAT (NP_000344) produced in HEK293T cell. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI3G6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 6898 | |
| IgG2a | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title