missing translation for 'onlineSavingsMsg'
Learn More

Tasquinimod, MedChemExpress

Produktkod. 16476737
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
16476737 5 mg 5mg
16455016 100 mg 100mg
16454028 10 mg 10mg
16410387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16476737 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY10528
 Restricted Item
Estimated Shipment: 29-09-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Tasquinimod is an oral antiangiogenic agent, which has the potential for castration-resistant prostate cancer treatment. Tasquinimod binds to the regulatory Zn2+ binding domain of HDAC4 with Kd of 10-30 nM. Tasquinimod also is a S100A9 inhibitor.

Tasquinimod is an oral antiangiogenic agent, which has the potential for castration-resistant prostate cancer treatment. Tasquinimod binds to the regulatory Zn2+ binding domain of HDAC4 with Kd of 10-30 nM. Tasquinimod also is a S100A9 inhibitor.

Specifikationer

Chemical Name or Material Tasquinimod
CAS 254964-60-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H17F3N2O4
Quantity 5 mg
Synonym ABR-215050
Solubility Information DMSO : ≥ 42 mg/mL (103.36 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(C1=C(O)C2=C(N(C)C1=O)C=CC=C2OC)N(C)C3=CC=C(C(F)(F)F)C=C3
Molecular Weight (g/mol) 406.36
Formula Weight 406.36
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.