missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TASP1 Antibody (OTI1H6), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74444R
This item is not returnable.
View return policy
Description
TASP1 Monoclonal specifically detects TASP1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| TASP1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TASP1 | |
| Full length recombinant protein of human TASP1 (NP_060184) produced in HEK293T cells. | |
| 0.1 mL | |
| 55617.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1H6 | |
| Western Blot | |
| C20orf13taspase 1, chromosome 20 open reading frame 13, dJ585I14.2, EC 3.4.25.-, FLJ20212, MGC39159, taspase, threonine aspartase, 1, Taspase-1, threonine aspartase 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction