missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TASP 0390325

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TASP 0390325 is a potent and selective V1B receptor antagonist (IC50 = 2.22 nM for rat pituitary receptor). Exhibits no significant affinity for 85 other receptors, ion channels and transporters at 10 μM, including V1A and V2. Increases ACTH levels in rats. Orally bioavailable. Antidepressant and anxiolytic.
Specifications
Specifications
| CAS | 1642187-96-9 |
| Quantity | 10 mg |
| Target | Vasopressin Receptor |
| Molecular Formula | C25H29ClFN5O4.HCl |
| Formula Weight | 554.44g/mol |
| Product Type | Antagonist |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction