missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TASP 0390325
GREENER_CHOICE

Product Code. 18720118
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18720118

Brand: Tocris Bioscience™ 5729/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective V1B receptor antagonist; orally bioavailable

TASP 0390325 is a potent and selective V1B receptor antagonist (IC50 = 2.22 nM for rat pituitary receptor). Exhibits no significant affinity for 85 other receptors, ion channels and transporters at 10 μM, including V1A and V2. Increases ACTH levels in rats. Orally bioavailable. Antidepressant and anxiolytic.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1642187-96-9
Quantity 50 mg
Target Vasopressin Receptor
Molecular Formula C25H29ClFN5O4.HCl
Formula Weight 554.44g/mol
Product Type Antagonist

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.