missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TASK-5/KCNK15 Polyclonal specifically detects TASK-5/KCNK15 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | TASK-5/KCNK15 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | Acid-sensitive potassium channel protein TASK-5, dJ781B1.1, K2p15.1, KCNK11, KCNK14, KIAA0237, KT3.3, potassium channel subfamily K member 15, potassium channel, subfamily K, member 14, potassium channel, subfamily K, member 15, TASK-5, TASK5Two pore potassium channel KT3.3, TWIK-related acid-sensitive K(+) channel 5, Two pore K(+) channel KT3.3 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human TASK-5/KCNK15 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?