missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS6417, MedChemExpress
Description
TAS6417 (CLN-081) is a highly effective, orally active and pan-mutation-selective EGFR tyrosine kinase inhibitor with a unique scaffold fitting into the ATP-binding site of the EGFR hinge region, with IC50 values ranging from 1.1-8.0 nM.
Specifications
Specifications
| Chemical Name or Material | TAS6417 |
| CAS | 1661854-97-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H20N6O |
| Quantity | 100 mg |
| Synonym | CLN-081 |
| Solubility Information | DMSO : 22.73 mg/mL (57.34 mM; Need ultrasonic) |
| SMILES | NC1=C2C(N3C(C(C)=C[C@@H](C3)NC(C=C)=O)=C2C4=CC5=CC=CC=C5N=C4)=NC=N1 |
| Molecular Weight (g/mol) | 396.44 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?