missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R60 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556466
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TSLGRHRKKA LLTTSGFREP SVQAHIKA Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 54%.
Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.Specifications
TAS2R60 | |
Polyclonal | |
Unconjugated | |
TAS2R60 | |
MGC119154; MGC119155; T2R56; T2R60; TAS2R60; taste 2 receptor member 60; taste receptor type 2 member 56; Taste receptor type 2 member 60; taste receptor, type 2, member 60 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
338398 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P59551 | |
TAS2R60 | |
Recombinant protein corresponding to Human TAS2R60 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only