missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS2R60 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599930
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TAS2R60.
Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.
Specifications
| TAS2R60 | |
| Polyclonal | |
| Unconjugated | |
| TAS2R60 | |
| MGC119154; MGC119155; T2R56; T2R60; TAS2R60; taste 2 receptor member 60; taste receptor type 2 member 56; Taste receptor type 2 member 60; taste receptor, type 2, member 60 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 338398 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P59551 | |
| TAS2R60 | |
| A synthesized peptide derived from human TAS2R60(Accession P59551), corresponding to amino acid residues M204-M254. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido