missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R45 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5102300
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TAS2R45.
Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. [UniProt]
Specifications
| TAS2R45 | |
| Polyclonal | |
| Unconjugated | |
| TAS2R45 | |
| GPR59; G-protein coupled receptor 59; MGC119923; MGC119924; MGC119925; T2R45; TAS2R45; taste 2 receptor member 45; taste receptor type 2 member 45; taste receptor, type 2, member 45; ZG24P | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 259291 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P59539 | |
| TAS2R45 | |
| A synthesized peptide derived from human TAS2R45(Accession P59539), corresponding to amino acid residues Q229-L279. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction