missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TAS2R43 Polyclonal specifically detects TAS2R43 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | TAS2R43 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | T2R43, T2R52Taste receptor type 2 member 52, taste receptor type 2 member 43, taste receptor, type 2, member 43 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human TAS2R43 (NP_795365.2). Peptide sequence MLANLVPFTVTLISFLLLVCSLCKHLKKMQLHGKGSQDPSTKVHIKVLQT |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?