missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS2R41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563394
This item is not returnable.
View return policy
Description
Immunogen sequence: NFYYSAQKVE YSGGLGRQFF H Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 52%.
TAS2R41 is a receptor that may play a role in the perception of bitterness and belongs to the G-protein receptor superfamily. TAS2R41 is expressed mainly in taste receptor cells of the tongue and the palate eptithelia. Structurally, the TAS2R41 gene encodes a seven trans-membrane receptor protein and is clustered together with eight other taste receptor genes on chromosome 7. The compound chloramphenicol is an agonist for the TAS2R41 protein. Further, TAS2R41 may play a role in sensing the chemical composition of the gastrointestinal content. The activity of TAS2R41 may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. TAS2R41 is expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.
Specifications
| TAS2R41 | |
| Polyclonal | |
| Unconjugated | |
| TAS2R41 | |
| MGC119928; MGC119929; mGR26; mt2r35; T2R12; T2R26; T2R41; T2R59; Tas2r12; Tas2r126; Tas2r26; Tas2r41; taste 2 receptor member 41; taste receptor rT2R12; taste receptor type 2 member 41; Taste receptor type 2 member 59; taste receptor, type 2, member 126; taste receptor, type 2, member 41 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 259287, 266656 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P59536 | |
| TAS2R12P, TAS2R41 | |
| Recombinant protein corresponding to Human TAS2R41. Recombinant protein control fragment (Product #RP-101629). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction