missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS2R41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567769
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
TAS2R41 is a receptor that may play a role in the perception of bitterness and belongs to the G-protein receptor superfamily. TAS2R41 is expressed mainly in taste receptor cells of the tongue and the palate eptithelia. Structurally, the TAS2R41 gene encodes a seven trans-membrane receptor protein and is clustered together with eight other taste receptor genes on chromosome 7. The compound chloramphenicol is an agonist for the TAS2R41 protein. Further, TAS2R41 may play a role in sensing the chemical composition of the gastrointestinal content. The activity of TAS2R41 may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. TAS2R41 is expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.
Spécification
| TAS2R41 | |
| Polyclonal | |
| Unconjugated | |
| TAS2R41 | |
| MGC119928; MGC119929; mGR26; mt2r35; T2R12; T2R26; T2R41; T2R59; Tas2r12; Tas2r126; Tas2r26; Tas2r41; taste 2 receptor member 41; taste receptor rT2R12; taste receptor type 2 member 41; Taste receptor type 2 member 59; taste receptor, type 2, member 126; taste receptor, type 2, member 41 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 246219, 259287, 266656, 387353 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P59532, P59536, Q9JKE7 | |
| Tas2r126, TAS2R12P, TAS2R41 | |
| Synthesized peptide derived from human TAS2R41. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu