missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS2R30 Polyclonal Antibody

Description
Antibody detects endogenous levels of total TAS2R47.
Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. [UniProt].
Specifications
Specifications
| Antigen | TAS2R30 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TAS2R30 |
| Gene Accession No. | P59541 |
| Gene Alias | T2R30; T2R44; T2R47; T2R53; TAS2R30; TAS2R44; TAS2R47; taste 2 receptor member 30; taste receptor T2R47; taste receptor type 2 member 30; Taste receptor type 2 member 47; taste receptor, type 2, member 30; taste receptor, type 2, member 47; type 2 taste receptor member 30; type 2 taste receptor member 47 |
| Gene Symbols | TAS2R30 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?