missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5102254
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TAS2R49.
The sense of taste provides animals with valuable information about the quality and nutritional value of food. A family of G protein-coupled receptors are involved in taste perception and include T1R, which is involved in sweet and umami taste perception and T2R, which is involved in bitter taste perception. Both types of taste receptors couple to various G proteins to initiate signal transduction cascades. T2R49 plays a role in sensing the chemical composition of the gastrointestinal content. T2R49 is expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells.
Specifications
| TAS2R20 | |
| Polyclonal | |
| Unconjugated | |
| TAS2R20 | |
| MGC119118; MGC119119; MGC119120; MGC119121; MGC134637; T2R20; T2R49; T2R56; TAS2R20; TAS2R49; taste 2 receptor member 20; taste receptor type 2 member 20; Taste receptor type 2 member 49; taste receptor type 2 member 56; taste receptor, type 2, member 20; taste receptor, type 2, member 49 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 259295 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P59543 | |
| TAS2R20 | |
| A synthesized peptide derived from human TAS2R20(Accession P59543), corresponding to amino acid residues L98-H148. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction