Learn More
Invitrogen™ TAS2R19 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5102253
Product Code. 16678724
  • £457.00 / 100µL
Description
Antibody detects endogenous levels of total TAS2R48.
Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TAS2R19
Polyclonal
Unconjugated
TAS2R19
MGC126853; MGC138203; MSTP058; T2R19; T2R23; T2R48; TAS2R19; TAS2R23; TAS2R48; taste 2 receptor member 19; taste receptor type 2 member 19; Taste receptor type 2 member 23; Taste receptor type 2 member 48; taste receptor, type 2, member 19; taste receptor, type 2, member 23; taste receptor, type 2, member 48
Rabbit
Affinity chromatography
RUO
259294
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P59542
TAS2R19
A synthesized peptide derived from human TAS2R19(Accession P59542), corresponding to amino acid residues L185-L235.