Learn More
Invitrogen™ TAS2R14 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5102250
Product Code. 16648724
  • £449.00 / 100µL
Description
Antibody detects endogenous levels of total TAS2R14.
This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13. Gustducin-coupled receptor implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in subsets of taste receptor cells of the tongue and palate epithelium and exclusively in gustducin-positive cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TAS2R14
Polyclonal
Unconjugated
TAS2R14
MGC125491; MGC125492; T2R14; TAS2R14; taste 2 receptor member 14; taste receptor family B member 1; Taste receptor type 2 member 14; taste receptor, family B, member 1; taste receptor, type 2, member 14; TRB1
Rabbit
Affinity chromatography
RUO
50840
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NYV8
TAS2R14
A synthesized peptide derived from human TAS2R14(Accession Q9NYV8), corresponding to amino acid residues K231-I281.