missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS2R13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102249
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TAS2R13.
This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13. Gustducin-coupled receptor implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in subsets of taste receptor cells of the tongue and palate epithelium and exclusively in gustducin-positive cells.
Specifications
| TAS2R13 | |
| Polyclonal | |
| Unconjugated | |
| TAS2R13 | |
| T2R13; TAS2R13; taste 2 receptor member 13; taste receptor family B member 3; Taste receptor type 2 member 13; taste receptor, family B, member 3; taste receptor, type 2, member 13; TRB3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 50838 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NYV9 | |
| TAS2R13 | |
| A synthesized peptide derived from human TAS2R13(Accession Q9NYV9), corresponding to amino acid residues L138-L188. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction