missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00 - £251.00
Specifications
| Antigen | TAS2R1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637527
|
Novus Biologicals
NBP3-12625-100ul |
100 μg |
£251.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18630629
|
Novus Biologicals
NBP3-12625-50ul |
50 μg |
£143.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAS2R1 Polyclonal antibody specifically detects TAS2R1 in Human samples. It is validated for Western BlotSpecifications
| TAS2R1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS, pH7.0 | |
| 50834 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC126778, T2R1MGC126780, Taste receptor family B member 7, taste receptor, family B, member 7, taste receptor, type 2, member 1, TRB7taste receptor type 2 member 1 | |
| Produced in rabbits immunized with E. coli-derived Human TAS2R1 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title