missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS1R3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534255
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Baboon (100%) Orangutan (89%).
The protein encoded by T1R3 is a G-protein coupled receptor involved in taste responses. The encoded protein can form a heterodimeric receptor with TAS1R1 to elicit the umami taste response, or it can bind with TAS1R2 to form a receptor for the sweet taste response.
Specifications
| TAS1R3 | |
| Polyclonal | |
| Unconjugated | |
| TAS1R3 | |
| Sac; Saccharin preference protein; sweet taste receptor T1R3; T1r3; TAS1R3; taste 1 receptor member 3; taste receptor type 1 member 3; taste receptor, type 1, member 3; TR3; TS1R3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83756 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q7RTX0 | |
| TAS1R3 | |
| Synthetic 19 amino acid peptide from N-terminal extracellular domain of human TAS1R3. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction