missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS1R3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93271-0.1ml
This item is not returnable.
View return policy
Description
TAS1R3 Polyclonal antibody specifically detects TAS1R3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TAS1R3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| T1R3Sweet taste receptor T1R3, taste receptor type 1 member 3, taste receptor, type 1, member 3, TR3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 400-570 of human TAS1R3 (NP_689414.2). VAQALHNTLQCNASGCPAQDPVKPWQLLENMYNLTFHVGGLPLRFDSSGNVDMEYDLKLWVWQGSVPRLHDVGRFNGSLRTERLKIRWHTSDNQKPVSRCSRQCQEGQVRRVKGFHSCCYDCVDCEAGSYRQNPDDIACTFCGQDEWSPERSTRCFRRRSRFLAWGEPAVL | |
| 0.1 mL | |
| GPCR | |
| 83756 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction