missing translation for 'onlineSavingsMsg'
Learn More

TAS1R2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSR00183W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11585063

  • £393.00 / 100µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μL of sterile water.

Putative taste receptor. TAS1R2/TAS1R3 recognizes diverse natural and synthetic sweeteners. Expressed mainly in circumvallate and foliate taste papillae.
Specifications

Specifications

TAS1R2
Polyclonal
Tas1r2
Taste receptor type 1 member 2 (precursor), sweet taste receptor T1R2, G-protein coupled receptor 71, Tas1r2, Gpr71, T1r2, Tr2, TS1R2
Rabbit
100 μL
Primary
Mouse
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q925I4
TAS1R2
Synthetic peptide from mouse taste receptor, type 1, member 2.
RUO
83770
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers