missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS1R2 Polyclonal Antibody

Description
Predicted to react with Mouse and Rat samples.
Putative taste receptor. TAS1R2/TAS1R3 recognizes diverse natural and synthetic sweeteners. Expressed mainly in circumvallate and foliate taste papillae.
Specifications
Specifications
| Antigen | TAS1R2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TAS1R2 |
| Gene Accession No. | Q8TE23, Q925I4, Q9Z0R7 |
| Gene Alias | aldehyde dehydrogenase 4 family, member A1; Aldh4a1; candidate taste receptor T1R2; G protein-coupled receptor 71; GPCRB4; GPR71; G-protein coupled receptor 71; Sweet taste receptor T1R2; T1R2; Tas1r2; taste 1 receptor member 2; taste receptor TR2; taste receptor type 1 member 2; taste receptor, type 1, member 2; Tr2; TS1R2 |
| Gene Symbols | TAS1R2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?