missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS1R1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534253
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bovine, Cat, Dog, Elephant, Panda, Horse, Pig (100%).
Putative taste receptor. TAS1R1/TAS1R3 responds to the umami taste stimulus (the taste of monosodium glutamate) and also to most of the 20 standard L-amino acids, but not to their D-enantiomers or other compounds. Sequence differences within and between species can significantly influence the selectivity and specificity of taste responses. Expressed strongly only in fungiform papillae.
Specifications
| TAS1R1 | |
| Polyclonal | |
| Unconjugated | |
| TAS1R1 | |
| candidate taste receptor T1R1; G protein-coupled receptor 70; GM148; GPCRB3; GPCR-B3; GPR70; G-protein coupled receptor 70; seven transmembrane helix receptor; T1r1; TAS1R1; taste 1 receptor member 1; taste receptor type 1 member 1; taste receptor, type 1, member 1; TR1; TS1R1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100052182, 100626842, 489633, 523473, 80835 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q7RTX1 | |
| TAS1R1 | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human TAS1R1. | |
| 50 μg | |
| Primary | |
| Human, Canine, Bovine, Pig, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction