missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tartrate Resistant Acid Phosphatase Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584356
This item is not returnable.
View return policy
Description
Immunogen sequence: GGVPNAPFHT AREMANAKEI ARTVQILGAD FILSLGDNFY FTGVQDINDK RFQETFEDVF SDRSLRKVPW YVLAGNHDHL GNVSAQI Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 90%.
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.
Specifications
| Tartrate Resistant Acid Phosphatase | |
| Polyclonal | |
| Unconjugated | |
| ACP5 | |
| acid phosphatase 5, tartrate resistant; acid phosphatase type V; Acp5; HPAP; human purple acid phosphatase; T5ap; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; tartrate-resistant acid phosphatase type 5; TRACP; TRACP5a; TRACP5b; TRAP; TR-AP; TRAPase; TrATPase; TTRRAP; Type 5 acid phosphatase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P13686 | |
| ACP5 | |
| Recombinant protein corresponding to Human ACP5. Recombinant protein control fragment (Product #RP-103307). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction