missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tartaric Acid NF/FCC Fine Granular, Macron Fine Chemicals™
Specifications
Specifications
| CAS | 87-69-4 |
| Molecular Formula | C4H6O6 |
| MDL Number | MFCD00064207 |
| Synonym | l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid |
| InChI Key | FEWJPZIEWOKRBE-UHFFFAOYNA-N |
| SMILES | OC(C(O)C(O)=O)C(O)=O |
| IUPAC Name | 2,3-dihydroxybutanedioic acid |
| Molecular Weight (g/mol) | 150.09 |
| PubChem CID | 444305 |
| ChEBI | CHEBI:15671 |
| Show More |
Safety and Handling
Product Identifier
- L(+)-Tartaric Acid
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: L(+)-Tartaric acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?