missing translation for 'onlineSavingsMsg'
Learn More

(+)-Tartaric Acid, Extra Pure, SLR, Crystals, Fisher Chemical™

Product Code. 11483653 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483653 250 g 250g
10306010 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483653 Supplier Fisher Chemical Supplier No. T/0200/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 168°C
Color White
pH 1.6
Packaging Plastic powder jar
Quantity 250 g
Formula Weight 150.09g/mol
Vapor Pressure <0.1mbar at 20°C
Physical Form Solid
compliance-icons
Product Identifier
  • L(+)-Tartaric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.