missing translation for 'onlineSavingsMsg'
Learn More

Tartaric acid disodium dihydrate, MedChemExpress

Product Code. 16471188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471188 Supplier Medchem Express Supplier No. HYD0850
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tartaric acid disodium dihydrate is a Acid phosphatase inhibitor, is a sodium salt used in buffers for molecular biology and cell culture applications.

Tartaric acid disodium dihydrate is a Acid phosphatase inhibitor, is a sodium salt used in buffers for molecular biology and cell culture applications.

Specifications

Chemical Name or Material Tartaric acid disodium dihydrate
CAS 6106-24-7
Color White
Purity Grade Notes Research
Molecular Formula C4H10Na2O8
Quantity 5 g
Synonym Sodium tartrate dibasic dihydrate Sodium tartrate dihydrate
Solubility Information H2O : 100 mg/mL (430.85 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES OC([C@H](O)[C@@H](O)C(O)=O)=O.[2H2O].[2Na]
Molecular Weight (g/mol) 232.1
Formula Weight 232.1
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.