missing translation for 'onlineSavingsMsg'
Learn More

(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™

Product Code. 10614742 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10614742 1 kg 1kg
10060400 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10614742 Supplier Fisher Chemical Supplier No. T/0250/60
  • ONLINE EXCLUSIVE PRICE
    £137.00
    £112.34 / 1kg
    Save £24.66
    18% Off
 Restricted Item
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

CAS: 87-69-4 | Molecular Formula: C4H6O6 | Molecular Weight: 150.09 g/mol

(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 168°C
Color White
pH 1.6
Packaging Plastic powder jar
Quantity 1 kg
Formula Weight 150.09g/mol
Vapor Pressure <0.1mbar at 20°C
Physical Form Solid
compliance-icons
Product Identifier
  • L(+)-Tartaric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.