missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TARSH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £434.00
Specifications
| Antigen | TARSH |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229255
|
Novus Biologicals
NBP3-35446-20ul |
20 μL |
£154.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227782
|
Novus Biologicals
NBP3-35446-100ul |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TARSH Polyclonal antibody specifically detects TARSH in Human samples. It is validated for ELISA,Western BlotSpecifications
| TARSH | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ABI family, member 3 (NESH) binding protein, ABI gene family member 3-binding protein, DKFZP586L2024, FLJ41743, Nesh-binding protein, NeshBP, NESHBPABI gene family, member 3 (NESH) binding protein, target of Nesh-SH3, Tarsh, TARSHFLJ41754 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-280 of human TARSH (NP_056244.2).,, Sequence:, QKLPKGKRPNLKVHINTTSDSILLKFLRPSPNVKLEGLLLGYGSNVSPNQYFPLPAEGKFTEAIVDAEPKYLIVVRPAPPPSQKKSCSGKTRSRKPLQLVVGTLTPSSVFLSWGFLINPHHDWTLPSHCPNDRFYTIRYREKDKEKKWIFQICPATETIVENLKPNTVYEFGVKDNVEGGIWSKIFNHKTVVGSKKVNGKIQSTYDQDHTVPAYVPRKLIPITIIKQVIQNVTHKDSAKSPEKAPLGGVILV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 25890 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title