missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531649
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (88%), Rat (87%), Chimpanzee (100%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.
Specifications
| TARS2 | |
| Polyclonal | |
| Unconjugated | |
| TARS2 | |
| 2610024N01Rik; AI429208; COXPD21; RGD1308283; TARS2; Tarsl1; threonine--tRNA ligase, mitochondrial; Threonyl-tRNA synthetase; threonyl-tRNA synthetase 2, mitochondrial (putative); threonyl-tRNA synthetase, mitochondrial; threonyl-tRNA synthetase-like 1; ThrRS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80222 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.83 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BW92 | |
| TARS2 | |
| Recombinant protein encompassing a sequence within the center region of human TARS2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction